Learn More
BUCHI Multivapor™ P-12 Parallel Evaporator with Rotavapor Connection
Catalog No. 05405377
Change view
Click to view available options
Volume (Metric):
12 x 30 mL
Catalog No. Volume (Metric)
05405377 12 x 30 mL
1 options
Catalog No. 05405377 Supplier BUCHI Supplier No. MP12001R00
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A compact, 12-position parallel vortex evaporator that achieves high efficiency for multiple sample evaporation with an easy operation.
  • Increase efficiency by processing up to 12 samples in parallel
  • Full visibility of the samples during the evaporation process
  • All glass parts, except the sample vessels, are Plastic+Glas coated to provide maximum safety
  • Increase sample throughput with a fraction collector directly using the Multivapor
  • Connection piece included for condensor, but condensor is sold separately
  • Includes tube adapters, preparation/transfer rack, and 30 mL test tubes
Specifications
For Use With (Application) Parallel Evaporation
For Use With (Equipment) Rotary Evaporator (R-100 or R-300) and Associated Vacuum Controller
Type Parallel Evaporator
Volume (Metric) 12 x 30 mL
Includes Tube Adapters, Preparation/Transfer Rack, and 30 mL Test Tubes