missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MultiShot™ TOP10 Chemically Competent E. coli Cells, 96-tube rack

Catalog No. A40007049
Change view
Click to view available options
Quantity:
1 x 96 Tubes (50 μL/Well)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A40007049 1 x 96 Tubes (50 μL/Well)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A40007049 Supplier Invitrogen™ Supplier No. A40007049
Only null left

MultiShot TOP10 Chemically Competent E. coli Cells provide high-efficiency transformation for routine cloning with flexible workflow capacity. The 96-tube rack format supports scalable, high-throughput plasmid propagation and blue/white screening.

Invitrogen MultiShot TOP10 Chemically Competent E. coli Cells are derived from DH10B cells and are one of the most popular laboratory strains. Provided in a tube rack of 96 tubes to allow flexible use in high-throughput workflows and help achieve transformation efficiencies of >1 x 108 cfu/μg control plasmid DNA, Multishot TOP10 cells are a versatile strain that can be used in multiple routine cloning applications.

Features of MultiShot TOP10 competent cells include:

  • TOP10 strain ideal for high-efficiency cloning and plasmid propagation
  • Competent cells pre-aliquoted into a rack of 96 tubes, ready for transformation
  • mcrA mutation that supports efficient transfection of methylated DNA
  • lacZΔM15 allele that permits rapid blue/white screening of transfectants
  • endA1 mutation that helps reduce endonuclease activity and improve plasmid DNA yields
  • recA1 mutation that helps reduce unwanted recombination

MultiShot TOP10 competent cells are designed to facilitate high-throughput transformation. This format allows all steps of the transformation protocol, up to plating, to take place in the same well, saving time and reducing the chance of contamination. After the addition of DNA, MultiShot TOP10 competent cells can be transformed by heat-shock at 42°C using a heat block or thermocycler in the same plate.

Genotype

F-mcrA Δ(mrr-hsdRMS-mcrBC) Φ80lacZΔM15 ΔlacX74 recA1 araD139 Δ(ara, leu)7697 galU galK rpsL (StrR) endA1 nupG

Specifications

Product Type Chemically Competent Cells
Includes pUC19 DNA, S.O.C media
Transformation Efficiency Level ≥1.0 x 108
Shipping Condition Dry Ice
Species E. coli
Format Tube Rack
Product Line MultiShot
For Use With (Application) Cloning
Quantity 1 x 96 Tubes (50 μL/Well)

For Research Use Only.