Learn More
Invitrogen™ MultiShot™ Stbl3™Chemically Competent E. coli Cells, 96-tube rack
Description
Invitrogen MultiShot Stbl3 Chemically Competent E. coli Cells are designed for cloning direct repeats found in lentiviral expression vectors as these cells reduce the frequency of homologous recombination of direct repeats found in lentiviral vectors (ViraPower Lentiviral Expression kits) and other retroviral vectors (instability-prone HIV-based plasmids).
Features of MultiShot Stbl3 competent cells include:
- Expected efficiency of >1 x 108 transformants/μg plasmid DNA
- Optimal performance for the propagation of lentiviral vectors and maintenance of direct repeats
- Enhanced genomic DNA cloning capacity (mcrB, mrr, and hsdS20)
- Capable of producing high DNA yields
The Stbl3 strain has been essential for many applications, including genome editing CRISPR workflows, vaccine development (including SARS-CoV-2 vaccine research), cloning, and maintaining vectors with multiple repeats (e.g., gene coding protein with repeated sequence). The transformation efficiency of MultiShot Stbl3 chemically competent cells is >1 x 108 cfu/μg plasmid DNA.
The Stbl3 strain is derived from the HB101 E. coli strain with a hybrid K-12 and B strain genome. It contains the recA13 allele of the recA mutation that reduces nonspecific recombination occurrence in cloned DNA. Mutations in the Stbl3 methylation-dependent restriction system (mcrB, mrr, and hsdS20) allow construction of cloning genomic DNA. This strain has endA1+, a nonspecific endonuclease I that digests DNA. The Stbl3 strain also reproducibly yields more plasmid DNA than DH strains (10-fold or more plasmid DNA than the TOP10 strain) and sustainably produces high titer lentivirus.
MultiShot Stbl3 competent cells offer:
- Expected efficiency of >1 x 108 transformants/μg plasmid DNA
- Optimal performance for the propagation of lentiviral vectors and maintenance of direct repeats
- Enhanced genomic DNA cloning capacity (mcrB, mrr, and hsdS20)
- Capable of producing high DNA yields
Note: For optimal performance and to maintain plasmid with complex repeat sequences, Stbl3 cells should be cultured at 30°C. When performing plasmid DNA isolation from Stbl3 cells that have wild type endonuclease I (endA1+), ensure that buffers contain 10 mM EDTA. EDTA will inactivate the endonuclease and avoid DNA nicking and vector degradation (follow plasmid DNA purification kit instructions for endA1+ E. coli strains).
Genotype
F–mcrB mrr hsdS20(rB–, mB–) recA13 supE44 ara-14 galK2 lacY1 proA2 rpsL20(StrR) xyl-5 λ–leu mtl-1
Specifications
Specifications
| Product Type | Chemically Competent Cells |
| Includes | pUC19 DNA, S.O.C media |
| Transformation Efficiency Level | ≥1.0 x 107 |
| Shipping Condition | Dry Ice |
| Species | E. coli |
| Format | Tube Rack |
| Product Line | MultiShot |
| For Use With (Application) | Cloning |
| Quantity | 1 x 96 Tubes (50 μL/Well) |
For Research Use Only.