Learn More
Invitrogen™ MultiShot™ DH5α™ T1R Chemically Competent E. coli Cells, 96-tube rack
Description
Invitrogen MultiShot DH5α T1R Chemically Competent E. coli Cells are a well-known, versatile strain that is used in many everyday cloning applications.
Features of DH5α T1R competent cells include:
- TOP10 strain ideal for high-efficiency cloning and plasmid propagation
- Microtiter plate format design convenient for automated high-throughput cloning
- mcrA mutation that supports efficient transfection of methylated DNA
- lacZΔM15 allele that permits rapid blue/white screening of transfectants
- endA1 mutation that helps reduce endonuclease activity and improve plasmid DNA yields
- recA1 mutation that helps reduce unwanted recombination
MultiShot DH5α T1R competent cells are prepared by a proprietary modification of the Hanahan method. These cells are suitable for use in high efficiency transformations in applications where a low amount of DNA is used or to construct libraries using plasmid-derived vectors. In addition to supporting blue/white screening, recA1 and endA1 mutations in DH5α cells increase insert stability and improve the quality of plasmid DNA prepared from minipreps.
The Φ80lacZΔM15 marker provides α-complementation of the β-galactosidase gene from pUC or similar vectors and can be used for blue/white screening of colonies on bacterial plates containing Bluo-gal or X-gal. DH5α cells are capable of being transformed efficiently with large plasmids and can also serve as a host for the M13mp cloning vectors if a lawn of DH5αF'IQ (or similar) is provided to allow plaque formation.
Genotype
F– φ80lacZΔ M15 Δ (lacZYA-argF) U169 recA1 endA1 hsdR17 (rK– mK+) phoA supE44 λ- thi–1 gyrA96 relA1
Specifications
Specifications
| Product Type | Chemically Competent Cells |
| Includes | pUC19 DNA, S.O.C media |
| Transformation Efficiency Level | ≥1.0 x 108 |
| Shipping Condition | Dry Ice |
| Species | E. coli |
| Format | Tube Rack |
| Product Line | MultiShot |
| For Use With (Application) | Cloning |
| Quantity | 1 x 96 Tubes (50 μL/Well) |
For Research Use Only.