missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ MultiRack Reload Tip System

Catalog No. 07202412
Change view
Click to view available options
Volume (Metric):
10 μL
200 μL
300 μL
1250 μL
Tip Style:
Extended Length
Universal
Rack Color:
Blue
Green
Red
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Tip Style Rack Color
07202412 10 μL Extended Length Red
07202413 200 μL Universal Yellow
07202414 300 μL Universal Green
07202415 1250 μL Extended Length Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 07202412 Supplier Axygen™ Supplier No. MR10XTRL

MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in Bulk, Racked, Filter and Reload packaging styles to meet the variety of needs in the lab setting.

The Axygen MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in bulk, racked, filter, and reload packaging styles to meet the variety of needs in the lab setting. The MultiRack tip system offers standard and extended length tips covering volumes from 1 μL to 1250 μL. For racked and reload packaging styles, the tips are pre-loaded into easy-load color-coded deck inserts. With the smaller footprint and reusable rack, this system uses 30% less plastic, minimizing the environmental impact. The stackable design saves critical bench space. Axygen MultiRack Tip System products are made in the USA and meet ISO standards. All tips are free of detectable RNase, DNase, DNA, and pyrogens. For more details visit our Certificates page at www.corning.com/lifesciences.
Features
• Reusable rack with reload systems 
• Smaller footprint and stackable design, saving valuable bench space
• Extended tips offered in 10, 300, and 1250 μL (1000 μL extended length) volumes
• Color-coded racks for easy volume identification

 

Specifications

Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Non-sterile
Color Red
Rack Color Red
Autoclavable Autoclavable
Model MR-10XT-RL
Certifications/Compliance ISO 13845
Clean Claims Free of detectable DNAse/RNAse, human DNA, and PCR inhibition
For Use With (Application) Diagnostics
Format Refill
Tip Style Extended Length
Packaging Reload System
Quantity 960/pk, 4 pks/cs
Product Type Axygen MultiRack Reload System
Includes Tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line MultiRack
Material Polyethylene
Show More Show Less