missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Multiplexed Proteomics™ Phosphoprotein Gel Stain Kits (with Pro-Q™ Diamond and SYPRO™ Ruby Gel Stains)

Catalog No. M33305
Encompass
Change view
Click to view available options
Quantity:
1 Set, 1 L per Dye
1 Set, 200 mL per Dye
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M33305 1 Set, 1 L per Dye
M33306 1 Set, 200 mL per Dye
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M33305 Supplier Invitrogen™ Supplier No. M33305

Our new Pro-Q Diamond Phosphoprotein Gel Stain provides a convenient method for selectively staining phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein-specific antibodies and western blot analysis.

Our new Pro-Q Diamond Phosphoprotein Gel Stain provides a convenient method for selectively staining phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein-specific antibodies and western blot analysis. This reagent, when combined with our total protein stain, SYPRO Ruby Protein Gel Stain, becomes a powerful combination for quantitative proteome analysis.

Our new Pro-Q Diamond Phosphoprotein Gel Stain provides a convenient method for selectively staining phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein-specific antibodies and western blot analysis. This reagent, when combined with our total protein stain, SYPRO Ruby Protein Gel Stain, becomes a powerful combination for quantitative proteome analysis. This convenient Pro-Q Diamond Phosphoprotein Gel Stain and SYPRO Ruby Protein Gel Stain Pack provides each stain in 1-liter amounts (Cat. No. M-33305) or in 200-mL amounts (Cat. No. M-33306).

Features include:

  • Sensitive—Pro-Q Diamond Phosphoprotein Gel Stain offers detection level down to 1–16 ng of phosphoprotein per band
  • Convenient—selectively stain phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein-specific antibodies and western blot analysis. SYPRO Ruby Gel Stain is subsequently used for total protein staining and simplifies the determination of phosphorylation level
  • Linear signal—quantify with ease as Pro-Q Diamond Phosphoprotein Gel Stain and SYPRO Ruby Gel Stain exhibit excellent linearity over three orders of magnitude, ideal for quantitation

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage • Pro-Q Diamond Phosphoprotein Gel Stain, 1 L
• SYPRO Ruby Protein Gel Stain, 1 L.

Store at room temperature and protect from light.
Description Multiplexed Proteomics™ Phosphoprotein Gel Stain Kit #1
Detection Location In-Gel Detection
Detection Method Fluorescence
Label or Dye SYPRO Ruby, Pro-Q Diamond
Quantity 1 Set, 1 L per Dye
Shipping Condition Room Temperature
Target Molecule Proteins (Phosphoproteins)
Product Line PRO-Q, SYPRO
Product Type Phosphoprotein Gel Stain Kit