Learn More
United Scientific™ Multimeter
Description
- Compact analog multimeter offers popular features such as AC voltage, DC voltage, DC current, Ohms, measures capacitance and decibels, and comes with overload protection and diode test.
- Includes test leads and instruction manual
- Operates on 2 AA size batteries (Batteries not included)
Specifications
Specifications
| For Use With (Application) | Classroom Demonstrations |
| Product Type | Multimeter |
| Display | Analog |
| Quantity | 1 Ea. |
Warning: Cancer and reproductive harm