missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek MultiFlo FX Multi-Mode Dispenser PROMO

MultiFlo FX is an automated multi-mode reagent dispenser for 6- to 1536-well microplates.

Specifications

Product Type Microplate Dispenser
Accuracy ±5% at 1 μL, ±2.0% at 5 μL, ±2.0% at 10 μL
Certifications/Compliance CE and TUV marked, RoHS compliant
For Use With (Application) Primary/secondary screening assays, compound storage, genomics and proteomics research, ELISA, cell based washing, fixing, staining, volume or concentration normalization
For Use With (Equipment) BioStack and third party automation compatible, BioSpa 8 Automated Incubator compatible
View More Specs

Products 3
Catalog Number Mfr. No. Model Includes Price Quantity & Availability  
Catalog Number Mfr. No. Model Includes Price Quantity & Availability  
BTMFXP1SIP
View Documents Promotion Details
Agilent BioTek
MFXP1SI3096
MFXP1 1 module arm, 1 peristaltic dispense pump and 1 peri-pump cassette with plastic tips N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTMFXP2RP
View Documents Promotion Details
Agilent BioTek
MFXP2RSN3096
MFXP2 2 module arms, 1 peristaltic dispense pump and 1 peri-pump cassette with plastic tips N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTMFXP2SIP
View Documents Promotion Details
Agilent BioTek
MFXP2SI3096
MFXP2R RAD single channel dispenser, 2 module arms, 2 peristaltic dispense pumps, 1 RAD manifold and tubing N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

MultiFlo FX is an automated multi-mode reagent dispenser for 6- to 1536-well microplates. The system incorporates several unique technologies in its modular design, such as Parallel Dispense, random access dispense (RAD) and patented Automated Media Exchange (AMX) modules

The technologies in MultiFlo FX microplate dispenser enable many liquid handling applications from 2D and 3D cell culture to concentration normalization assays, ELISA, bead-based assays and more. A fully configured MultiFlo FX replaces up to five liquid handlers, saving space, time, and instrumentation budgets. MultiFlo FX delivers complete workflow automation in cell imaging and biochemical applications.

Specifications

Specifications

Microplate Dispenser
CE and TUV marked, RoHS compliant
BioStack and third party automation compatible, BioSpa 8 Automated Incubator compatible
Dispensing (peri pump and syringe): 96-, 384- and 1536-well standard, deep and PCR plates; 6-. 12-, 24- and 48-well plates (dispense tip configurable); Washing: 96-. 384-well standard plates; 6-, 12-, 24-,and 48-well plates (with compatible manifold configuration)
LHC2 Software, LHC2 Secure for 21 CFR Part 11 compliance (option), SiLA Compliant driver (option)
17.2 in.
8 in.
11.8 in.
14.3 lb.
90 W max.
±5% at 1 μL, ±2.0% at 5 μL, ±2.0% at 10 μL
Primary/secondary screening assays, compound storage, genomics and proteomics research, ELISA, cell based washing, fixing, staining, volume or concentration normalization
Color Touchscreen
<5% CV at 1 μL, <2.5% CV at 5 μL, <2.0% CV at 10 μL
96 wells, 5 μL cass, 10 μL/well: 3 sec., 96 wells, 5 μL cass, 20 μL/well: 3.5 sec., 384 wells, 5 μL cass, 5 μL/well: 6.5 sec., 384 wells, 10 μL cass, 10 μL/well: 8 sec., 384 wells, 1 μL cass, 1 μL/well: 6 sec., 1536 wells, 1 μL cass, 1 μL/well: 21 s
43.5 cm
20.3 cm
29.2 cm
6.5 kg
100/240 V
Videos
SDS
Documents

Documents