missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek MultiFlo FX Multi-Mode Dispenser, IVD PROMO

Catalog No. BTMFXP2RSIP
Change view
Click to view available options
Model:
MFXP1
MFXP2
MFXP2R
Includes:
1 module arm, 1 peristaltic dispense pump and 1 peri-pump cassette with plastic tips
2 module arms, 1 peristaltic dispense pump and 1 peri-pump cassette with plastic tips
RAD single channel dispenser, 2 module arms, 2 peristaltic dispense pumps, 1 RAD manifold and tubing
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Includes
BTMFXP2RSIP MFXP2R RAD single channel dispenser, 2 module arms, 2 peristaltic dispense pumps, 1 RAD manifold and tubing
BTMFXP1P MFXP1 1 module arm, 1 peristaltic dispense pump and 1 peri-pump cassette with plastic tips
BTMFXP2P MFXP2 2 module arms, 1 peristaltic dispense pump and 1 peri-pump cassette with plastic tips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BTMFXP2RSIP Supplier Agilent BioTek Supplier No. MFXP2RSI3096

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MultiFlo FX is an automated multi-mode reagent dispenser for 6- to 1536-well microplates. For In Vitro Diagnostic Use.

MultiFlo FX is an automated multi-mode reagent dispenser for 6- to 1536-well microplates. The system incorporates several unique technologies in its modular design, such as Parallel Dispense, random access dispense (RAD) and patented Automated Media Exchange (AMX) modules

The technologies in MultiFlo FX microplate dispenser enable many liquid handling applications from 2D and 3D cell culture to concentration normalization assays, ELISA, bead-based assays and more. A fully configured MultiFlo FX replaces up to five liquid handlers, saving space, time, and instrumentation budgets. MultiFlo FX delivers complete workflow automation in cell imaging and biochemical applications.

Specifications

Accuracy ±5% at 1 μL, ±2.0% at 5 μL, ±2.0% at 10 μL
Product Type Microplate Dispenser
Certifications/Compliance CE and TUV marked, RoHS compliant
Volume (Metric) 500 nL to 30,000 μL
For Use With (Application) Primary/secondary screening assays, compound storage, genomics and proteomics research, ELISA, cell based washing, fixing, staining, volume or concentration normalization
For Use With (Equipment) BioStack and third party automation compatible, BioSpa 8 Automated Incubator compatible
Includes RAD single channel dispenser, 2 module arms, 2 peristaltic dispense pumps, 1 RAD manifold and tubing
Interface Color Touchscreen
Model MFXP2R
Plate Type Dispensing (peri pump and syringe): 96-, 384- and 1536-well standard, deep and PCR plates; 6-. 12-, 24- and 48-well plates (dispense tip configurable); Washing: 96-. 384-well standard plates; 6-, 12-, 24-,and 48-well plates (with compatible manifold configuration)
Precision <5% CV at 1 μL, <2.5% CV at 5 μL, <2.0% CV at 10 μL
Software LHC2 Software, LHC2 Secure for 21 CFR Part 11 compliance (option), SiLA Compliant driver (option)
Speed 96 wells, 5 μL cass, 10 μL/well: 3 sec., 96 wells, 5 μL cass, 20 μL/well: 3.5 sec., 384 wells, 5 μL cass, 5 μL/well: 6.5 sec., 384 wells, 10 μL cass, 10 μL/well: 8 sec., 384 wells, 1 μL cass, 1 μL/well: 6 sec., 1536 wells, 1 μL cass, 1 μL/well: 21 s
Width (English) 17.2 in.
Width (Metric) 43.5 cm
Height (English) 8 in.
Height (Metric) 20.3 cm
Depth (English) 11.8 in.
Depth (Metric) 29.2 cm
Weight (English) 14.3 lb.
Weight (Metric) 6.5 kg
Electrical Requirements 90 W max.
Voltage 100/240 V
Show More Show Less