Learn More
Agilent BioTek MultiFlo FX Multi-Mode Dispenser, IVD

MultiFlo FX is an automated multi-mode reagent dispenser for 6- to 1536-well microplates. For In Vitro Diagnostic Use.
Supplier: Agilent BioTek MFXP2RSI
Description
MultiFlo FX is an automated multi-mode reagent dispenser for 6- to 1536-well microplates. The system incorporates several unique technologies in its modular design, such as Parallel Dispense, random access dispense (RAD) and patented Automated Media Exchange (AMX) modules
The technologies in MultiFlo FX microplate dispenser enable many liquid handling applications from 2D and 3D cell culture to concentration normalization assays, ELISA, bead-based assays and more. A fully configured MultiFlo FX replaces up to five liquid handlers, saving space, time, and instrumentation budgets. MultiFlo FX delivers complete workflow automation in cell imaging and biochemical applications.
Chemical Identifiers
| Microplate Dispenser | |
| CE and TUV marked. RoHS compliant. | |
| BioStack and third party automation compatible; BioSpa 8 Automated Incubator compatible | |
| Dispensing (peri pump and syringe): 96-, 384- and 1536-well standard, deep and PCR plates; 6-. 12-, 24- and 48-well plates (dispense tip configurable); Washing: 96-. 384-well standard plates; 6-, 12-, 24-,and 48-well plates (with compatible manifold configuration) | |
| LHC2 Software; LHC2 Secure for 21 CFR Part 11 compliance (option); SiLA Compliant driver (option) | |
| 17.2 in. | |
| 8 in. | |
| 11.8 in. | |
| 14.3 lb. | |
| 90 W max. |
| ±5% at 1 μL, ±2.0% at 5 μL, ±2.0% at 10 μL | |
| Primary/secondary screening assays, compound storage, genomics and proteomics research, ELISA, cell based washing, fixing, staining, volume or concentration normalization | |
| Color touch screen; Create, edit or run multiple protocols onboard | |
| <5% CV at 1 μL, <2.5% CV at 5 μL, <2.0% CV at 10 μL | |
| 96 wells, 5 μL cass, 10 μL/ well: 3 s, 96 wells, 5 μL cass, 20 μL/ well: 3.5 s, 384 wells, 5 μL cass, 5 μL/ well: 6.5 s, 384 wells, 10 μL cass, 10 μL/ well: 8 s, 384 wells, 1 μL cass, 1 μL/ well: 6 s, 1536 wells, 1 μL cass, 1 μL/ well: 21 s | |
| 43.5 cm | |
| 20.3 cm | |
| 29.2 cm | |
| 6.5 kg | |
| 100/240 V |
Specifications
| ±5% at 1 μL, ±2.0% at 5 μL, ±2.0% at 10 μL | |
| CE and TUV marked. RoHS compliant. | |
| Primary/secondary screening assays, compound storage, genomics and proteomics research, ELISA, cell based washing, fixing, staining, volume or concentration normalization | |
| RAD single channel dispenser, 2 module arms, 2 peristaltic dispense pumps, 1 RAD manifold and tubing | |
| MFXP2R | |
| <5% CV at 1 μL, <2.5% CV at 5 μL, <2.0% CV at 10 μL | |
| 96 wells, 5 μL cass, 10 μL/ well: 3 s, 96 wells, 5 μL cass, 20 μL/ well: 3.5 s, 384 wells, 5 μL cass, 5 μL/ well: 6.5 s, 384 wells, 10 μL cass, 10 μL/ well: 8 s, 384 wells, 1 μL cass, 1 μL/ well: 6 s, 1536 wells, 1 μL cass, 1 μL/ well: 21 s | |
| 43.5 cm | |
| 20.3 cm | |
| 29.2 cm | |
| 6.5 kg | |
| 100/240 V |
| Microplate Dispenser | |
| 500 nL to 30,000 μL | |
| BioStack and third party automation compatible; BioSpa 8 Automated Incubator compatible | |
| Color touch screen; Create, edit or run multiple protocols onboard | |
| Dispensing (peri pump and syringe): 96-, 384- and 1536-well standard, deep and PCR plates; 6-. 12-, 24- and 48-well plates (dispense tip configurable); Washing: 96-. 384-well standard plates; 6-, 12-, 24-,and 48-well plates (with compatible manifold configuration) | |
| LHC2 Software; LHC2 Secure for 21 CFR Part 11 compliance (option); SiLA Compliant driver (option) | |
| 17.2 in. | |
| 8 in. | |
| 11.8 in. | |
| 14.3 lb. | |
| 90 W max. |