Learn More
Agilent BioTek MultiFlo FX Microplate Dispenser Accessory, Reagent Bottle
Provides supply containment
Supplier:  Agilent BioTek 7140511
Catalog No. BT7140511
Only null left
Explore available promotions
Description
Description
  • Autoclavable
  • With cap and connector
Specifications
Specifications
Reagent Bottle
CE and TUV marked. RoHS compliant.
Cap, connector
Autoclavable
For microfill, multiflo fx, precision, bio-tek dispensers and pipetting systems with autoclavable fluid paths
1 L
Product Suggestions
Product Suggestions