Learn More
Alkali Scientific Multi-Seal™ Multilayer Polyethylene Sealing Film, 50/pk
Catalog No. 502388853
Encompass
Multi-Seal™ Multilayer Polyethylene Sealing Film consisting of a top pierceable clear polyethylene film and a white polypropylene sublayer with 96 circular areas void of adhesive. The white sublayer aids in the alignment of the film to the plate; alignment of opposing corner wells ensures that all 96 wells will be properly aligned. The adhesive-free zones are 9mm on center to facilitate multi-channel pipetting without adhesive fouling of pipet tips or robotic probes. Offers excellent chemical resistance and is pierccable. Samples have minimal contact with adhesive due to the adhesive-free zones.
Catalog No. 502388853 Supplier Alkali Scientific Supplier No. SF36ML
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Multi-Seal™ Multilayer Polyethylene Sealing Film consisting of a top pierceable clear polyethylene film and a white polypropylene sublayer with 96 circular areas void of adhesive. The white sublayer aids in the alignment of the film to the plate; alignment of opposing corner wells ensures that all 96 wells will be properly aligned. The adhesive-free zones are 9mm on center to facilitate multi-channel pipetting without adhesive fouling of pipet tips or robotic probes. Offers excellent chemical resistance and is pierccable. Samples have minimal contact with adhesive due to the adhesive-free zones.