missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Multi-Purpose Sample Cups

Catalog No. 22170167
Change view
Click to view available options
Capacity (Metric):
0.25 mL
0.5 mL
1.5 mL
2 mL
4 mL
Quantity:
1000/Pk.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Quantity
22170167 2 mL 1000/Pk.
21200250 1.5 mL 1000/Pk.
22170251 0.5 mL 1000/Pk.
22170161 0.25 mL 1000/Pk.
22170166 4 mL 1000/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 22170167 Supplier Globe Scientific Supplier No. 110621
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For laboratory analyzers

  • Available in a variety of sizes and designed for use on most popular instruments in the industry
  • Polystyrene (PS) sample cups made to the exact specifications of the OEM
  • Provide excellent optical properties

Specifications

Quantity 1000/Pk.
Capacity (Metric) 2 mL
Product Type Sample Cup
Capacity (English) 0.06 oz.
Material Polystyrene
For Use With (Application) For use on the most popular instruments in the industry
For Use With (Equipment) Hitachi™ and many other laboratory analyzers