missing translation for 'onlineSavingsMsg'
Learn More

3M™ Multi-Purpose Duct Tape

Catalog No. 19072067
Change view
Click to view available options
Color:
Silver
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19072067 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19072067 Supplier 3M™ Supplier No. 7100029108

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

3M™ Multi-Purpose Duct Tape is constructed of polyethylene film laminated to cloth with a rubber adhesive to resist curling.

  • Designed for general use in MRO/construction applications
  • Tears off the roll cleanly for easy application
  • Construction allows for permanent and temporary applications.
  • Sticks immediately and stays on
  • Strong water-resistant backing resists wear and light abrasion
  • Conforms to irregular surfaces without lifting
  • Available in a wide variety of colors

Specifications

Color Silver
Material Rubber
Product Type Multi-purpose Duct Tape
For Use With (Application) Design and Construction, Manufacturing, Mining, Oil and Gas, Transportation
Model 3900
Quantity 24/Cs.
Thickness (English) 8.1 mil
Dimensions (L x W) 54.8 m x 48 mm