missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Multi-Position Stirrer

Catalog No. 01918310
Change view
Click to view available options
Stirring Positions:
4
9
Controller Type:
Analog
Digital
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stirring Positions Controller Type
01918310 9 Digital
01918307 4 Analog
01918308 9 Analog
01918309 4 Digital
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01918310 Supplier OHAUS™ Supplier No. 30573804

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use the Multi-Position Stirrer for synchronized and uniform mixing.

  • Applications: Dissolution Studies, Titrations, Media/Reagent Preparations, Synchronized Stirring of multiple samples
  • Display: Independent LED displays for speed and time (on digital models); Knob with 1-10 dial markings (on non-digital models)
  • Operation: N/A
  • Communication: N/A
  • Construction: Cast aluminum housing, glass filled nylon top plate. Includes stir bars to accommodate each stirring position
  • Design Features: Speed ramping feature slowly increases motor speed for enhanced coupling and improved stirring. Microprocessor-controlled for precise speed regulation. Timer counts up or when programmed to a set time will turn off stirring and sound alarm when timer reaches zero.

Specifications

Speed 60 to 1400 rpm
Stirring Positions 9
Top Plate Material Glass-filled Nylon
Capacity 250 mL
Display Digital LED
Shape Square
Voltage 120 V
Wattage 75 W
Height (English) 4 in.
Height (Metric) 970 mm
Length (English) 17 in.
Length (Metric) 432 mm
Top Plate Length (English) 12.5 in.
Top Plate Length (Metric) 318 mm
Top Plate Width (English) 11 in.
Top Plate Width (Metric) 279 mm
Width (English) 11 in.
Width (Metric) 279 mm
Amperage 1.3 A
Frequency 50/60 Hz
Certifications/Compliance UL, cUL
Controller Type Digital
Model STMP9DG
Temperature Range (Metric) 5°C to 40°C, 80% RH, Non-condensing
Timer 1 sec. to 160 hr.
Type Multi-Position Stirrers
Weight (English) 14 lb.
Weight (Metric) 6.3 kg
Show More Show Less