missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Multi Function 3D Rotators
For continuous operation, timed operation or different combinations to ensure optimal mixing conditions for any application
$1,684.06
Specifications
| Orbit | 23 mm |
|---|---|
| Rotating Angle | 0 to 360° |
| Load Bearing Capacity (Metric) | 1 kg |
| Speed Range | 5 to 100 rpm |
| Voltage | 100/240 V |
Includes: Dimpled rubber mat; 6-tube securing bands
| Catalog Number | Mfr. No. | Plug Type | Certifications/Compliance | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Plug Type | Certifications/Compliance | Price | Quantity & Availability | ||||
|
88861047
|
Fisherbrand™
88861047 |
US Plug | cCSAus |
Each for $1,684.06
|
|
||||
Description
- Rotation speed: 5 to 100 rpm for long wave motion
- Reciprocal rotation: 0°to 360° turning angle for long and medium way motion
- Vibration: 0° to 5° turning angle, programmable in bursts of 0 to 5 seconds, for short wave motion
- Fits neatly into small workspaces
- Operating temperature range of 5 to 40°C allows for use in refrigerators or non-CO2 incubators
- Continuous or timed operation with automatic switch-off
- Display will show last used settings, even after power has been turned off.
- Continue to run by last set speed and the remaining time when the power recovered.
- Audible and visual alarm: In timed mode, when the time reaches zero, alarm will sound 5 times, then the time window shows“End“ and flickers all the time. Alarm can be stopped by touch any key.
Warranty
2 years
Specifications
| 23 mm | |
| 1 kg | |
| 100/240 V | |
| 2.2 lb. | |
| 266 mm | |
|
| 0 to 360° | |
| 5 to 100 rpm | |
| 5°C to 40°C | |
| 306 mm | |
| Immune precipitations, Affinity matrix , Trypsinisation, Gel straining and distaining, Antibody staining, Washes, Hybridizations, Southern blots, Western blots | |
| 2 Years |