Learn More
Thermo Scientific™ Multi-cell accessories for GENESYS™ 140/150/180 and BioMate™ 160 Spectrophotometers
Description
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirements. Cable-free, snap-in design allows for easy exchange of accessories.
Speed up your analysis with high-throughput measurements.
Specifications
Specifications
| Compatibility | Accommodates four long-path rectangular cells up to 50mm. Please note: Not compatible with low volume cells (< 3.5mL) or any other type of cell holder. |
| Type | Rect Cell 4-position Carousel |
| For Use With (Equipment) | GENESYS G5 |
| Holds | 4 x 50mm PL Rectangular Cells |