Learn More
Contec Multi Bucket System with Flat Mops
Contec's Multi-Bucket System offers flexibility with two or three buckets in stainless steel or polypropylene.
$4,804.92 - $7,952.40
| Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
|
19991014
|
Contec
2650U2PF |
Polypropylene Bucket, Stainless Steel Cart |
Case for $4,804.92
|
|
|||||
|
19991016
|
Contec
2707U2SF |
Stainless Steel |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
The Contec Multi-Bucket System is designed for use with Contec's flat mops, offering flexibility, mobility, and easy customization for cleanroom cleaning.
- Configurable with two or three buckets in stainless steel or polypropylene
- Extendable front frame accommodates an additional bucket for added capacity
- Quiet, locking casters ensure smooth movement and stability
- Utility handle for easy transport of cleaning supplies
Hands-free mop removal tool available for VertiKlean™ MAX™ and QuickTask™ mop heads
- Integrated mop holder with brace for secure storage during transport
- Compatible with stainless steel downpress wringer for flat mops
- Autoclavable at 121°C for 30 minutes for sterilization in critical environments
Specifications
| 2 Bucket System |