Learn More
Mullite
CAS: 1302-93-8 | Al6O13Si2 | 426.05 g/mol
$120.04 - $252.44
Chemical Identifiers
| CAS | 1302-93-8 |
|---|---|
| Molecular Formula | Al6O13Si2 |
| Molecular Weight (g/mol) | 426.05 |
| MDL Number | MFCD00058866 |
| InChI Key | KZHJGOXRZJKJNY-UHFFFAOYSA-N |
| Synonym | Aluminum oxide/silicon oxide |
Description
Mullite is present in the form of needles in porcelain, it is used in high-mullite based products is in hot blast stove checker bricks, melting and firing processes, and is used as a refractory material, manufacture of a type of crucible, steel industry, electronic substrates and protective coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Safety and Handling
EINECSNumber : 215-113-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only