missing translation for 'onlineSavingsMsg'
Learn More

Mullite

Catalog No. AA2288122
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2288122 100 g
AA2288136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA2288122 Supplier Thermo Scientific Chemicals Supplier No. 02288122

CAS: 1302-93-8 | Al6O13Si2 | 426.05 g/mol

Mullite is present in the form of needles in porcelain, it is used in high-mullite based products is in hot blast stove checker bricks, melting and firing processes, and is used as a refractory material, manufacture of a type of crucible, steel industry, electronic substrates and protective coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1302-93-8
Molecular Formula Al6O13Si2
Molecular Weight (g/mol) 426.05
MDL Number MFCD00058866
InChI Key KZHJGOXRZJKJNY-UHFFFAOYSA-N
Synonym Aluminum oxide/silicon oxide

Specifications

Color White
Physical Form Powder
Quantity 100 g
Solubility Information Insoluble in water.
Odor Odorless
Chemical Name or Material Mullite
EINECSNumber 215-113-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only