Learn More
Thermo Scientific™ Mueller Hinton Agar
Catalog No. p-4524601
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R454082 500 g
1 options
Catalog No. R454082 Supplier Thermo Scientific™ Supplier No. R454082
Only null left
The Thermo Scientific™ Remel Mueller Hinton Agar is used for antimicrobial disk diffusion susceptibility testing.
The Thermo Scientific™ Remel Mueller Hinton Agar is used for antimicrobial disk diffusion susceptibility testing.
Specifications
Certifications/Compliance Conforms to CLSI standards. Industrial Reference: BAM, COMPF.
Description Mueller Hinton Agar
Form Powder
Product Type Agar
Quantity 500 g