missing translation for 'onlineSavingsMsg'
Learn More

Mucochloric Acid 98.0+%, TCI America™

Catalog No. M047225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M047225G 25 g
M0472500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M047225G Supplier TCI America Supplier No. M047225G

Chemical Identifiers

CAS 87-56-9
Molecular Formula C4H2Cl2O3
Molecular Weight (g/mol) 168.96
MDL Number MFCD00006966,MFCD00006966
InChI Key LUMLZKVIXLWTCI-IHWYPQMZSA-N
Synonym mucochloric acid, 2,3-dichloromaleic aldehyde acid, kyselina mukochlorova, 2z-2,3-dichloro-4-oxobut-2-enoic acid, 2-butenoic acid, 2,3-dichloro-4-oxo-, 2z, aldehydodichloromaleic acid, dichloromalealdehydic acid, unii-5i5877jhiw, kyselina mukochlorova czech, ccris 6597
PubChem CID 2771871
IUPAC Name (2Z)-2,3-dichloro-4-oxobut-2-enoic acid
SMILES OC(=O)C(\Cl)=C(\Cl)C=O

Specifications

Melting Point 127°C
Color White-Yellow
Quantity 25 g
UN Number 2923
Formula Weight 168.96
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Mucochloric Acid
EINECSNumber (2)-1164&(2)-1166&(2)-1502
RTECSNumber AS7700000
TSCA Yes
Recommended Storage Refrigerator