missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mucobromic Acid 99.0+%, TCI America™
$100.62 - $1393.59
Chemical Identifiers
| CAS | 488-11-9 |
|---|---|
| Molecular Formula | C4HBr2O3- |
| Molecular Weight (g/mol) | 256.857 |
| MDL Number | MFCD00063745 |
| InChI Key | NCNYEGJDGNOYJX-IHWYPQMZSA-M |
| Synonym | mucobromic acid, z-2,3-dibromo-4-oxo-2-butenoate, z-2,3-dibromo-4-oxobut-2-enoate, 2z-2,3-dibromo-4-oxobut-2-enoate, z-2,3-bis bromanyl-4-oxidanylidene-but-2-enoate |
| PubChem CID | 6994897 |
| IUPAC Name | (Z)-2,3-dibromo-4-oxobut-2-enoate |
| SMILES | C(=O)C(=C(C(=O)[O-])Br)Br |
Chemical Identifiers
| 488-11-9 | |
| 256.857 | |
| NCNYEGJDGNOYJX-IHWYPQMZSA-M | |
| 6994897 | |
| C(=O)C(=C(C(=O)[O-])Br)Br |
| C4HBr2O3- | |
| MFCD00063745 | |
| mucobromic acid, z-2,3-dibromo-4-oxo-2-butenoate, z-2,3-dibromo-4-oxobut-2-enoate, 2z-2,3-dibromo-4-oxobut-2-enoate, z-2,3-bis bromanyl-4-oxidanylidene-but-2-enoate | |
| (Z)-2,3-dibromo-4-oxobut-2-enoate |
Specifications
| 488-11-9 | |
| White | |
| MFCD00063745 | |
| 3261 | |
| NCNYEGJDGNOYJX-IHWYPQMZSA-M | |
| (Z)-2,3-dibromo-4-oxobut-2-enoate | |
| 6994897 | |
| ≥99.0% (T) | |
| Mucobromic Acid |
| 124°C | |
| C4HBr2O3- | |
| 25 g | |
| mucobromic acid, z-2,3-dibromo-4-oxo-2-butenoate, z-2,3-dibromo-4-oxobut-2-enoate, 2z-2,3-dibromo-4-oxobut-2-enoate, z-2,3-bis bromanyl-4-oxidanylidene-but-2-enoate | |
| C(=O)C(=C(C(=O)[O-])Br)Br | |
| 256.857 | |
| 257.87 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-1165
TSCA : Yes