missing translation for 'onlineSavingsMsg'
Learn More

Mucobromic acid, 98+%

CAS: 488-11-9 | C4HBr2O3- | 256.857 g/mol

Supplier:  Thermo Scientific Chemicals A1599836

 View more versions of this product

Catalog No. AAA1599836


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Mucobromic acid
121°C to 124°C
MFCD00063745
UN3261
mucobromic acid, z-2,3-dibromo-4-oxo-2-butenoate, z-2,3-dibromo-4-oxobut-2-enoate, 2z-2,3-dibromo-4-oxobut-2-enoate, z-2,3-bis bromanyl-4-oxidanylidene-but-2-enoate
C(=O)C(=C(C(=O)[O-])Br)Br
256.857
257.88
488-11-9
C4HBr2O3-
500 g
1705707
NCNYEGJDGNOYJX-IHWYPQMZSA-M
(Z)-2,3-dibromo-4-oxobut-2-enoate
6994897
≥98%

Chemical Identifiers

488-11-9
256.857
NCNYEGJDGNOYJX-IHWYPQMZSA-M
6994897
C(=O)C(=C(C(=O)[O-])Br)Br
C4HBr2O3-
MFCD00063745
mucobromic acid, z-2,3-dibromo-4-oxo-2-butenoate, z-2,3-dibromo-4-oxobut-2-enoate, 2z-2,3-dibromo-4-oxobut-2-enoate, z-2,3-bis bromanyl-4-oxidanylidene-but-2-enoate
(Z)-2,3-dibromo-4-oxobut-2-enoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 207-670-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only