missing translation for 'onlineSavingsMsg'
Learn More

Mucobromic acid, 98+%

Catalog No. AAA1599822
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1599822 100 g
AAA1599836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1599822 Supplier Thermo Scientific Chemicals Supplier No. A1599822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 488-11-9 | C4HBr2O3- | 256.857 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 488-11-9
Molecular Formula C4HBr2O3-
Molecular Weight (g/mol) 256.857
MDL Number MFCD00063745
InChI Key NCNYEGJDGNOYJX-IHWYPQMZSA-M
Synonym mucobromic acid, z-2,3-dibromo-4-oxo-2-butenoate, z-2,3-dibromo-4-oxobut-2-enoate, 2z-2,3-dibromo-4-oxobut-2-enoate, z-2,3-bis bromanyl-4-oxidanylidene-but-2-enoate
PubChem CID 6994897
IUPAC Name (Z)-2,3-dibromo-4-oxobut-2-enoate
SMILES C(=O)C(=C(C(=O)[O-])Br)Br

Specifications

Melting Point 121°C to 124°C
Quantity 100 g
UN Number UN3261
Beilstein 1705707
Formula Weight 257.88
Percent Purity ≥98%
Chemical Name or Material Mucobromic acid
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 207-670-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only