MW 338.32 g/mol, Purity >99%. Fluorogenic substrate for -D-galactosidase, yielding a blue fluorescent solution upon cleavage. Potentially useful in differentiating species within the family of Enterobacteriaceae and in the localization of -galactosidase in plants. Fluorescence: ex 317 nm; em 374 nm (pH 9.1); ex 360 nm; em 449 nm (Reaction product).
The product is subject to the following: Abcam Restricted Use Statement