Learn More
Abcam Inc MTSSL, thiol-specific spin label, 50MG
Catalog No. 502705898
Encompass
MW 264.4 Da, Purity >98%. Highly reactive thiol-specific spin label. For protein cysteine residue labeling, in protein structure and protein dynamics determination and in protein-protein and protein-oligonucleotide interaction studies. Active in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502705898 Supplier Abcam Inc Supplier No. AB14430850MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 264.4 Da, Purity >98%. Highly reactive thiol-specific spin label. For protein cysteine residue labeling, in protein structure and protein dynamics determination and in protein-protein and protein-oligonucleotide interaction studies. Active in vitro.
The product is subject to the following: Abcam Restricted Use Statement