missing translation for 'onlineSavingsMsg'
Learn More

Pyrogallol, Crystal, Reagent, ACS, Spectrum™ Chemical

Catalog No. 18606011
Change view
Click to view available options
Quantity:
125 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606011 125 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18606011 Supplier Spectrum Chemical Mfg Cor Supplier No. P1460125GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

P1460, 87-66-1, C6H3(OH)3

Spectrum™ Chemical Pyrogallol, Crystal, Reagent, ACS , also known as Pyrogallic acid, is one of three isomeric benzenetriols that is used for oxygen absorption in gas analysis as well as in hair dying. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 87-66-1
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
MDL Number MFCD00002192
InChI Key WQGWDDDVZFFDIG-UHFFFAOYSA-N
IUPAC Name benzene-1,2,3-triol
SMILES OC1=CC=CC(O)=C1O

Specifications

CAS Max % 1
Melting Point 131°C to 135°C
Boiling Point 309°C (588.2°F)
Ignition Residue 5e-005
Packaging Amber Glass Bottle
Quantity 125 g
Grade Reagent
Physical Form Crystals or Flakes
Chemical Name or Material Pyrogallol