missing translation for 'onlineSavingsMsg'
Learn More

Brady™ MSDS Binder

Catalog No. 19103211
Change view
Click to view available options
Width (Metric):
269.24 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (Metric)
19103211 269.24 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19103211 Supplier Brady™ Supplier No. 2023VF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Package includes binder only (information center and accessories sold separately)

Specifications

Format Binder
Includes (10) Training Booklets; MSDS Binder
Diameter (English) Ring 1.5 in.
Width (Metric) 269.24 mm
Height (English) 11 in.
Quantity Each
Depth (English) 2 in.
Material Polyethylene
Color Blue on White
Languages English
For Use With (Application) Hazard Communication
Show More Show Less