Learn More
MSA™ V-Gard™ Slotted Protective Cap with 1-Touch Suspension
Catalog No. 191303449
Change view
Click to view available options
Color:
Yellow
Catalog No. Color
191303449 Yellow
1 options
Not specifically designed to protect against lateral blows from the front, side or rear.
Catalog No. 191303449 Supplier MSA™ Supplier No. 10057443
Not specifically designed to protect against lateral blows from the front, side or rear.
Lightweight for all-day comfort
  • Polyethylene shell and suspension system work together as a protection system
  • Available in three sizes: small (6 to 71/8), standard (61/2 to 8), and large (71/2 to 81/2)
  • 1-Touch suspension allows one-handed slide adjustment and easy squeeze opening
Order Info
Contact your Fisher Scientific™ Customer Safety Representative for ordering information for sizes not shown.
Certifications
SEI certified.
Compliance
Meets or exceeds all applicable requirements of ANSI Z89.1-2003 for Type I helmets, Class E and CSA Z94.1 2005,
Specifications
Certifications/Compliance Meets or exceeds ANSI Z89.1-2003; Type I; Class E and CSA Z94.1 2005; SEI certified
Color Yellow
Material Polyethylene
Size Standard
Suspension Type 1-Touch