Learn More
MSA™ V-Gard™ 500 Vented Protective Cap
Catalog No. 296351378
Change view
Click to view available options
Color:
Blue
Hi-vis Orange
Suspension Type:
4-Point Ratchet
6-Point Ratchet
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Suspension Type
296351378 Hi-vis Orange 6-Point Ratchet
296351365 Blue 4-Point Ratchet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 296351378 Supplier MSA™ Supplier No. 10034035
Cooling vents along the crown improve air circulation and comfort
  • Comfortable, lightweight design
  • Polyethylene shell features contours above the ear for easy integration of hearing protection
  • Glaregard™ underbrim reduces reflective glare
  • Available with four- or six-point Fas-Trac™ ratchet suspension with absorbent sweatband
Compliance
Third-party certified to meet ANSI/ISEA Z98.1, Type I, Class C requirements.
Specifications
Certifications/Compliance Meets or exceeds ANSI Z89.1-2003, Class C and CSA Z94.1-2005.
Color Hi-vis Orange
Suspension Type 6-Point Ratchet