missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSA™ Ultra-Twin™ Full Facepiece Respirators
Respirators provide a secure fit and strong protection while offering several options including an inturned lip for a comfortable seal against the face and a large chin cup for easy positioning.
Supplier: MSA™ 480259
Requires : Comfo Series cartridges and filters (see separate listings).
Description
- Five suspension headstraps and roller buckles secure the facepiece while minimizing hair entanglement
- Wide coated polycarbonate lens resists scratching and chemicals
- Available in Hycar™™ rubber and silicone
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Specifications
| NIOSH 42 CFR Part 84 | |
| Black | |
| Full Facepiece Respirator | |
| Silicone | |
| Comfo™ Series cartridges and filters | |
| Anti-scratch | |
| 480259 |
| Threaded | |
| Silicone | |
| Medium | |
| Utilities, general industry, welding | |
| Nosecup and speaking diaphragm | |
| Polycarbonate | |
| 5-point |