Learn More
MSA™ Ultra-Twin™ Full Facepiece Respirators
Respirators provide a secure fit and strong protection while offering several options including an inturned lip for a comfortable seal against the face and a large chin cup for easy positioning.
Supplier:  MSA™ 471286
 View more versions of this product
Catalog No. 18619W
Only null left
Explore available promotions
Requires : Comfo Series cartridges and filters (see separate listings).
Description
Description
  • Five suspension headstraps and roller buckles secure the facepiece while minimizing hair entanglement
  • Wide coated polycarbonate lens resists scratching and chemicals
  • Available in Hycar™™ rubber and silicone
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Specifications
Specifications
NIOSH 42 CFR Part 84
Black
Full Facepiece Respirator
Hycar™
Comfo™ Series cartridges and filters
Anti-scratch
471286
Threaded
Hycar Rubber
Medium
Utilities, general industry, welding
Nosecup and speaking diaphragm
Polycarbonate
5-point
Product Suggestions
Product Suggestions