missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSA™ Comfo™ and Comfo Classic™ Half-Mask Respirators
Description
Requires
Comfo Series cartridges and filters (see separate listings).
- Protection from exposure to vapors, gases, and particulates
- SoftFeel™ facepiece material increases softness
- Choose from Hycar™ rubber or silicone
- Face seal design provides tight, comfortable fit
- Ideal markets include utilities, oil & gas, construction and general industry
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Specifications
Specifications
| Certifications (NIOSH) | NIOSH 42 CFR Part 84 |
| Color | Black |
| Material | Hycar Rubber |
| Product Type | Half-Mask Respirator |
| Size | Small |
| Facepiece Material | Hycar™ |
| For Use With (Application) | Utilities, oil and gas, construction, general industry, welding |
| For Use With (Equipment) | Comfo™ Series cartridges and filters |
| Model | 808075 |