missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSA™ Comfo™ Cartridges
Particulate, chemical, and combination cartridges for use with Comfo Classic™, Comfo Elite™, Ultra-Twin™ and Ultra Elite™ respirators
$197.67 - $284.60
Specifications
| Certifications/Compliance | NIOSH 42CFR, Part 84 |
|---|---|
| Product Type | Comfo™ Respirator Cartridge |
| For Use With (Application) | Asbestos abatement, painting, sanding, grinding, welding |
| For Use With (Equipment) | Comfo classic™, ultra-twin™, ultra elite twin™ cartridge and duo-twin™ respirator and certain half mask and full face advantage™ respirator |
| Catalog Number | Mfr. No. | Cartridge Type | Color | Filter Efficiency | Filter Type | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Cartridge Type | Color | Filter Efficiency | Filter Type | Price | Quantity | ||
|
18947
|
MSA™
492790 |
Organic Vapor, Acid Gas, Ammonia, Methylamine, Formaldehyde, Hydrogen Fluoride Cartridge | Olive | 95% | N95, R951 |
Pack of 10 Each for $284.60
|
|
||
|
19084714
|
MSA™
464032 |
Acid Gas Cartridge | White | 95% | N95, R951 |
Pack of 10 Each for $197.67
|
|
||
Description
- Low-profile particulate and ShortStack™ combination cartridges feature a patented low-profile design that improves field of vision
- Special adapter also allows use with Advantage™ 200LS, Advantage 420, Advantage 4000 and Advantage 1000 respirators Sparkfoe™ particulate cartridge is suitable for use in applications with sparking hazard
Note: Do not exceed maximum use concentrations established by regulatory standards.
Certifications
All Comfo cartridges are approved under NIOSH 42CFR Part 84.
Specifications
| NIOSH 42CFR, Part 84 | |
| Comfo™ Respirator Cartridge | |
| Asbestos abatement, painting, sanding, grinding, welding | |
| Comfo classic™, ultra-twin™, ultra elite twin™ cartridge and duo-twin™ respirator and certain half mask and full face advantage™ respirator |