Learn More
MSA™ Bump Cap
Catalog No. p-322912
Change view
Click to view available options
Color:
White
Catalog No. Color
296351360 White
1 options
Catalog No. 296351360 Supplier MSA™ Supplier No. 10033652
Only null left
Protect your workers from minor bumps and lacerations
  • Size-adjustable; no replaceable parts
  • Ideal for workplaces such as meat packing, food processing, pest control, meter reading, and automotive repair
  • Perforated sides improves air circulation by allowing cross ventilation
  • Interior plastic liner includes foam pad for additional comfort
  • Standard version is molded from durable high-density polyethylene and is adjustable from size 6.5 to 8
NOTE: Bump caps do not meet the ANSI Z89.1-1997 standard, and are not designed to protect against impact of falling or glancing objects.
Specifications
Color White
Material HDPE
Size Standard
Suspension Type Adjustable