missing translation for 'onlineSavingsMsg'
Learn More

MSA™ Advantage™ 3200 Full Facepiece Respirator

Catalog No. 19041906A
Change view
Click to view available options
Size:
Small
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19041906A Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19041906A Supplier MSA™ Supplier No. 10028996

Requires

NIOSH-approved Advantage series cartridges and filters.

Full-facepiece respirators feature a large, optically clear lens and a soft silicone seal for an undistorted view and pressure free fit.

  • Dual-port has lower breathing resistance than traditional full-facepiece respirators
  • Soft, pliable silicone facemask and rubber head harness don quickly
  • Pre-adjustable straps ensure a consistent fit and tighter seal
  • Optically correct, wraparound facepiece lens provides a wide, distortion-free field of vision
  • Scratch-resistant lens coating
  • Fog-reducing standard nosecup

Certifications

NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.

Specifications

ANSI Standard ANSI Z87.1
Certifications/Compliance AS NZS 1716:2003
Certifications (EN 136) Class 2
Certifications (NIOSH) NIOSH 42 CFR Part 84
Connection Type Bayonet
Color Blue
Material Silicone
Product Type Full Facepiece Respirator
Size Small
Facepiece Material Silicone
For Use With (Application) Utilities, oil and gas, construction, general industry, search and rescue
Harness Material Rubber
Material (Lens) Scratch-resistant
Model 10028996
No. of Ports Dual
Weight (English) 1.15 lb.
Weight (Metric) 0.522 kg
Show More Show Less