missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Oxide, Powder, 99.9%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Crystalline Powder or Needles |
| Quantity | 500 g |
| Molecular Formula | Ag2O |
| Linear Formula | Ag2O |
| Synonym | Argentous oxide, Disilver oxide, Silver (I) oxide, Silver(1+) oxide |
| Solubility Information | Solubility in Water: 0.0017 g/100 mL Water |
| IUPAC Name | Silver(1+) oxide |
| Molecular Weight (g/mol) | 231.74 |
| PubChem CID | 9369 |
| Show More |
Safety and Handling
| Recommended Storage | Light Sensitive, Keep tightly closed in light-resistant containers |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.