missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ mRNA Vaccines Education Kit

Catalog No. S03079
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S03079 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03079 Supplier United Scientific™ Supplier No. MRNAKIT1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

mRNA Vaccines Education Kit is a unique kit allows students a hands on experience learning how mRNA vaccines work.

The world recently witnessed the most rapid vaccine production ever, achieving another milestone in medical science. In response to the novel SARS-CoV-2 virus and the CoVID-19 pandemic, a messenger ribonucleic acid (mRNA) vaccine was developed and administered to billions worldwide in less than a year. Prior vaccine production took five to ten years.

Just as the wheel was invented long before the automobile, mRNA vaccines were built upon much previous scientific knowledge, dating back to the days of Gregor Mendel and possibly before.

This mRNA kit is designed to aid in the teaching of how mRNA vaccines provide immunity against harmful viruses. A colorful animated slide show illustrates virus structure, viral entry into body cells and the immune response. A summary of how mRNA vaccines are produced and why they provide lasting immunity without causing sickness follows.

Understanding that students learn best when information is presented orally, visually, and tactilely; teachers are provided with animated slides to project on a wall or screen. Plastic models representing cells and key structures allow students to tactilely simulate immune response to a virus. Student assessment is evaluated via activities utilizing kit models and follow-up questions.

The kit contains no consumables and can be shared among teachers. The durability of the plastic models will provide for years of use.
  • Use hands-on models to investigate how the immune system reacts to vaccines
  • Learn about mRNA vaccines, viruses and the human immune system
  • Includes 35 custom plastic models
  • Includes downloadable teacher resources

Specifications

Grade 9 to 12
Class Size 1 Lab Group
Product Type Classroom Kit