missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ MQuant™ Total Hardness Test Strips

Catalog No. M1100460001
Change view
Click to view available options
Range:
0.1 to 30°d
4 to 26°e
6 to 31°e
Graduations:
0.1 to 30°d
4 to 26°e
6 to 31°e
Test Method:
EDTA
Phthaleinkomplexone
No. of Tests:
100
1000
25000
50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range Graduations Test Method No. of Tests
M1100460001 6 to 31°e 6 to 31°e EDTA 100
M1100320001 4 to 26°e 4 to 26°e EDTA 1000
M1100470013 6 to 31°e 6 to 31°e EDTA 25000
M1169970001 0.1 to 30°d 0.1 to 30°d Phthaleinkomplexone 50
M1100250001 4 to 26°e 4 to 26°e EDTA 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. M1100460001 Supplier MilliporeSigma™ Supplier No. 1.10046.0001

Includes

Tube containing 100 strips

To determine the total hardness of water

  • Small and easy to handle
  • Flexible and universal application
  • EDTA reaction method ideal for screening
  • Immediate result produced through on-site analysis
  • PET film and low reagent content
  • Easily disposable

1.10025.0001
Color-scale graduation: 4 - 5 - 9 - 18 - 26°e

1.10046.0001
Color-scale graduation: 6 - 13 - 19 - 25 - 31°e

Reclose the tube containing the test strips immediately after use.

Analytical Quality Assurance, Standard for Total Hardness

Specifications

For Use With (Application) Analytical quality assurance, to determine the total hardness of water
For Use With (Equipment) Colorimeter
Graduations 6 to 31°e
No. of Tests 100
Range 6 to 31°e
Storage Requirements 15°C to 25°C
Test Method EDTA
Type Test Strip