missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ MQuant™ Peroxide Test Strips

Catalog No. M1100810001
Change view
Click to view available options
Quantity:
100 Strips/Pk.
25 Strips/Pk.
Range:
0.5 to 25 mg/L H2O2
10 to 100 mg/L H2O2
100 to 1000 mg/L H2O2
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Range
M1100810001 100 Strips/Pk. 10 to 100 mg/L H2O2
M1100110001 100 Strips/Pk. 0.5 to 25 mg/L H2O2
M1103370001 100 Strips/Pk. 100 to 1000 mg/L H2O2
M1100110002 25 Strips/Pk. 0.5 to 25 mg/L H2O2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. M1100810001 Supplier MilliporeSigma™ Supplier No. 1.10081.0001
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Tube containing 100 strips

To check for presence or absence of residues of Peroxide

  • Small and easy to handle
  • Flexible and universal application
  • Enzymatic reaction method ideal for screening
  • Immediate result produced through on-site analysis
  • PET film and low reagent content
  • Easily disposable

1.10011.0001
Color-scale graduation: 0.5 - 2 - 5 - 10 - 25mg/L H2O2

1.10081.0001
Color-scale graduation: 1 - 3 - 10 - 30 - 100mg/L H2O2

1.10337.0001
Color-scale graduation: 100 - 200 - 400 - 600 - 800 - 1000mg/L H2O2

Reclose the tube containing the test strips immediately after use.

For food and water analysis

Specifications

For Use With (Application) For testing residual concentrations of disinfectants on equipment or the product itself
For Use With (Equipment) Colorimeter
Graduations 1-3-10-30-100
No. of Tests 100
Quantity 100 Strips/Pk.
Range 10 to 100 mg/L H2O2
Storage Requirements 2°C to 8°C
Test Method Enzymatic Reaction
Type Peroxide Test Strips