missing translation for 'onlineSavingsMsg'
Learn More

VAI Veltek 1700 Series MP Protective Sleeves

Catalog No. 19547242
Change view
Click to view available options
Cleanroom Class:
Class 100 (ISO 5)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cleanroom Class
19547242 Class 100 (ISO 5)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19547242 Supplier VAI Supplier No. 1700PSS1792

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Heavyweight, non-woven, microporous material is low in particulate shedding and suited for use where maximum protection is needed.

  • Heavyweight, non-woven and high quality microporous (MP) coated material
  • Suited for use where maximum protection to product or wearer is required
  • Liquid and chemical resistant

Specifications

Size Universal
Material Microporous Fabric
Color White
Sterility Sterile
Disposable Single-use
Closure Type Elastic Ends
Cuff Style Elastic
Model 1700
Includes Integrated Thumb Loop
Packaging Individually Packaged, Pair of Two
Cleanroom Class Class 100 (ISO 5)
Show More Show Less