missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ YNB, Powdered Media: Without Ammonium Sulfate

Catalog No. MP114027022
Change view
Click to view available options
Quantity:
227 g
454 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP114027022 454 g
MP114027012 227 g
MP114027032 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP114027022 Supplier MP Biomedicals™ Supplier No. 114027022

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Can be prepared by adding a carbon source, the appropriate amino acids, and agar if necessary

  • Nitrogen-based yeast

Yeast cell culture, Gene expression, Transcription regulation

Specifications

Certifications/Compliance Molecular Biology Certified™
Content And Storage 15°C to 30°C
Form Powder
Media Type YNB
Product Type Dehydrated Culture Media
Formulation Nitrogen Source: Ammonium Sulfate 5000mg/L; Vitamins: Biotin 0.002mg/L, Calcium Pantothenate 0.4mg/L, Folic Acid 0.002mg/L, Inositol 2mg/L, Niacin 0.4mg/L, P-Aminobenzoic Acid 0.2mg/L, Pyridoxine Hydrochloride 0.4mg/L, Riboflavin 0.2mg/L, Thiamine Hydrochloride 0.4mg; Trace Elements: Boric Acid 0.5mg, Copper Sulfate 0.04g, Potassium Iodide 0.1mg, Ferric Chloride 0.2mg, Manganese Sulfate 0.4mg, Sodium Molybdate 0.2mg, Zinc Sulfate 0.4mg; Salts: Magnesium Sulfate 500mg, Sodium Chloride 100mg, Potassium Phosphate monobasic 1000mg, Calcium Chloride 100mg
For Use With (Application) For vigorous growth of Saccharomyces cerevisiae
Quantity 454 g