missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Yeast, Torula, Powder
Source: Candida utilis
Supplier: MP Biomedicals™ 0290308580
Description
Applications:
Torula yeast is a common flavoring additive in processed foods such as chips, seasoning blends, and crackers. It is also used in pet foods, in beekeeping, and as an attractant in traps designed to control olive flies
Specifications
| Animal Diet | |
| Torula Yeast is a common flavoring additive in processed foods such as chips, seasoning blends and crackers, also used in pet foods, in beekeeping and as an attractant in traps designed to control olive flies | |
| 15°C to 30°C |
| Powder | |
| 45.35 kg |