missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Yeast, Torula, Powder
Source: Candida utilis
Manufacturer: MP Biomedicals™ 0290308505
Description
Applications:
Torula yeast is a common flavoring additive in processed foods such as chips, seasoning blends, and crackers. It is also used in pet foods, in beekeeping, and as an attractant in traps designed to control olive flies
Specifications
Animal Diet | |
Torula Yeast is a common flavoring additive in processed foods such as chips, seasoning blends and crackers, also used in pet foods, in beekeeping and as an attractant in traps designed to control olive flies | |
15°C to 30°C |
Powder | |
2.26 kg |