missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Yeast, Torula, Powder

Source: Candida utilis

Supplier:  MP Biomedicals™ 0290308525

 View more versions of this product

Catalog No. ICN90308525


Only null left

Description

Description

Applications:
Torula yeast is a common flavoring additive in processed foods such as chips, seasoning blends, and crackers. It is also used in pet foods, in beekeeping, and as an attractant in traps designed to control olive flies

Specifications

Chemical Identifiers

Animal Diet
Powder
Torula Yeast is a common flavoring additive in processed foods such as chips, seasoning blends and crackers, also used in pet foods, in beekeeping and as an attractant in traps designed to control olive flies
15°C to 30°C

Specifications

Animal Diet
Torula Yeast is a common flavoring additive in processed foods such as chips, seasoning blends and crackers, also used in pet foods, in beekeeping and as an attractant in traps designed to control olive flies
15°C to 30°C
Powder
11.33 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions