missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Williams' Medium E, powder, with L-glutamine

Catalog No. icn1050122
Change view
Click to view available options
Quantity:
1 x 10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN1050122 1 x 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN1050122 Supplier MP Biomedicals™ Supplier No. 091050122
Only null left

for the long-term cultivation of adult rat liver epithelial cells

Specifications

Product Type Williams medium E
Without Additives No Sodium Bicarbonate
Quantity 1 x 10 L
pH 6.8 to 7.3
Content And Storage 0° to 5°C