missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Wheat Germ Agglutinin, 5mg

Catalog No. ICN790162
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN790162 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN790162 Supplier MP Biomedicals™ Supplier No. 08790162

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

isolated from wheat germ, dissolved in 0.05 M Tris-HCl, 0.15 M NaCl buffer, pH 8.6 and purified by affinity chromatography

Specifications

Quantity 5 mg
Form Powder
For Use With (Application) Immunoassay
Product Type Wheat germ