missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Tryptose Phosphate Broth
Spécifications
Spécifications
| Content And Storage | 15°C to 30°C |
| Form | Liquid |
| Media Type | Tryptose Phosphate Broth |
| Product Type | Microbiology Media |
| Formulation | Tryptose 20g/L, Sodium Chloride 5g/L, Dextrose 2g/L, Disodium Phosphate 2.5g/L |
| Quantity | 100 mL |