missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Tryptic Soy Broth
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| Form | Powder |
| Media Type | Tryptic Soy Broth |
| Product Type | Microbiology Media |
| Formulation | Casein digest Peptone 17g/L, Soy Peptone 3g/L, Disodium Phosphate 2.5g/L, Dextrose 2.5g/L, Sodium Chloride 5g/L |
| For Use With (Application) | Isolation and cultivation of non-fastidious organisms |
| Quantity | 500 g |